Screw cap, DURAN® PURE GLS 80®
Leverandør: DWK Life Sciences
891169109
SCOT891169109EA 580 NOK
SCOT891169109
Screw cap, DURAN® PURE GLS 80®
Lokk Flaskelokk
The DURAN® PURE GLS 80® screw cap is made from PSU with highly inert PTFE coated silicone seal. It is designed for use with DURAN® PURE GLS 80® bottles.
  • Compatible with autoclaving and other common sterilization methods
  • With batch identification moulded into cap
  • Packaged in ISO Class 7 clean room
  • Single batch delivery
  • Certificate of Analysis and Change Control on request
Typical applications: Manufacturing of pharmaceutical products, storage of bulk drug intermediates and ingredients, transportation of APIs.