Embra® Optimia Non Woven Compress
Leverandør: Onemed Sverige AB
22300303
SELA22300303EA 94 NOK
SELA22300303
Embra® Optimia Non Woven Compress
Førstehjelpsutstyr Bandasjer
EMBRA® Optimia Nonwoven compress, 4-ply are produced from soft and highly absorptive nonwoven material. They are primarily used for cleaning and exudate absorption of wounds and abrasions.
  • 70% renewable materials (viscose)
  • High quality 70% viscose and 30% polyester, nonwoven material
  • Low linting and high softness
  • Keeps its shape, even when wet
  • Free from binding agents and optical brighteners
  • Effective in liquid and exudate absorption
  • Less adherent to the wound than gauze devices
Available in several sizes and can be used for external wound care,More Product Information