Drums, UN approved, with screw lids
Leverandør: SEMADENI
0719
SEMA0719EA 5200 NOK
SEMA0719
Drums, UN approved, with screw lids
Tønner, fat, trommer
Drums, made of blue HDPE.
  • Very sturdy
  • Good chemical resistance
  • Good temperature resistance
  • UN-approved