Container, PS, with Screw Cap
Leverandør: SEMADENI
10728
SEMA10728EA 680 NOK
SEMA10728
Container, PS, with Screw Cap
Prøvecontainere Beholdere, generelt bruk
Transparent, with flat bottom and tightly closing screw cap made of polyethylene.
Forpakning: Package contains 200 cans and lids individually packed in a bag.