Specimen Container, with Screw Cap
Leverandør: SEMADENI
11669
SEMA11669EA 3725 NOK
SEMA11669
Specimen Container, with Screw Cap
Prøvecontainere Beholdere, generelt bruk
Robust, transparent sample container made from polypropylene (PP) with tightly sealing yellow screw cap made from hard polyethylene (PE-HD). With easy to read engraved scale.
  • Autoclavable and reusable
  • Diameter: 58 mm
  • Graduation: 10 ml
Particularly suited for the storage and transportation of sample material from the laboratory and medicine.