Tubing clamps, POM
Leverandør: SEMADENI
2492
SEMA2492EA 2225 NOK
SEMA2492
Tubing clamps, POM
Klemmer Slangeklemmer
Polyacetal (POM), blue. Suitable for catheters, tubing and pipes up to 10 mm diameter.
  • Solid and sterilisable
  • Quick and easy opening and closing
  • Temperature resistant up to +130 °C