Jars with screw cap
Leverandør: SEMADENI
3046
SEMA3046EA 5825 NOK
SEMA3046
Jars with screw cap
Krukker Bokser, generell bruk
Jars, made of PP, with double walled bottom and tri-seal insert in the lid.