Desiccator cabinet, horizontal, PMMA
Leverandør: SEMADENI
3463
SEMA3463EA 52000 NOK
SEMA3463
Desiccator cabinet, horizontal, PMMA
Eksikatorer Eksikatorskap
Transparent PMMA cabinet that has white ABS frame and transparent PVC sides. Ideal for storing moisture sensitive samples or electronic components. Unit is not suitable for use under vacuum or pressure.
  • Door mounted hygrometer is graduated from 0 to 100%
  • Cabinet has two lateral interlocking gas connectors. Check valves open automatically and close when hose is disconnected, allowing gas to escape
Leveringsinformasjon: Supplied with 2 removable acrylic glass shelves.