Jars, wide mouth, with screw cap
Leverandør: SEMADENI
3742
SEMA3742EA 170 NOK
SEMA3742
Jars, wide mouth, with screw cap
Krukker Bokser, generell bruk
Jars, wide mouth, made of HDPE, white.
  • Space saving
  • With inner seal and red screw cap
  • Good chemical and temperature resistance