AssetTag
Leverandør: SenseAnywhere
0096647494982
SENA0096647494982EA 180 NOK
SENA0096647494982
AssetTag
Dataloggere
An AssetTag is a card with barcode for the holder and QR code for on the outside of an asset representing the monitored asset in SAClient.
  • Monitor temp/RH of the asset
  • Data loggers can be switched at any time
  • Complete temp/RH history of assets without any monitoring-gaps
  • No reconfiguration needed (alarms, reports etc.)
  • No paperwork or administration
  • Not prone to error or hassle
  • Complete audit trail
How it works: The back of the card has two-sided foam tape. The card can be snapped into two parts. Bottom part: attach to holder in the asset. Top part:More Product Information