AccessPoint Outdoor Active PoE
Leverandør: SenseAnywhere
0096647495132
SENA0096647495132EA 3225 NOK
SENA0096647495132
AccessPoint Outdoor Active PoE
Dataloggere
The SenseAnywhere AccessPoint is an economic and reliable plug and play device that connects SenseAnywhere’s wireless Sensors to the Cloud service of SenseAnywhere.
  • Wireless protocol – SenseAnywhere wireless protocol with automatic service discovery and seamless roaming
  • Wired protocol – 10Mbit Ethernet 10BASE-T, TCP/IP port 80, DHCP, DNS
  • Ethernet connector – Shielded RJ45
  • Compatibility – CE, EN300328, EN55022 (868 MHz)
  • FCC, IC (915 MHz)
  • IP65 protection
  • Number of data loggers supported – no limits
  • Power connector – DC plug, 5,5 / 2,1 mm
SenseAnywhere'sMore Product Information