Module for AiroSensor X ER 20-20-43, Digital PT100 4-wire Hermetically Sealed Temperature Probe
Leverandør: SenseAnywhere
0096647495163
SENA0096647495163EA 2000 NOK
SENA0096647495163
Module for AiroSensor X ER 20-20-43, Digital PT100 4-wire Hermetically Sealed Temperature Probe
Temperaturprober
This smart probe featuring a digital SenseAnywhere Bus interface has been especially designed for measuring extreme temperatures in liquid substances. Thanks to its hermetic seal made of PFA, better known as Teflon®, the Hermetically Sealed Pt100 Probe has a high level of chemical resistance. The probe is thanks to its PFA insulation waterproof and easy to clean. It can withstand aggressive substances, allowing it to take measurements in virtually all liquid fluids while being constantly submerged. The PT100 element has a Class A accuracy; measurements are provided at a very high accuracyMore Product Information