Module for AiroSensor 20-20-41, Magnetic Door Contact Set
Leverandør: SenseAnywhere
0096647495187
SENA0096647495187EA 410 NOK
SENA0096647495187
Module for AiroSensor 20-20-41, Magnetic Door Contact Set
Dataloggere
The SenseAnywhere AiroSensor 20-20-41 with Magnetic Door Contact Set is the ultimate solution to your refrigeration and freezer challenges!
  • Upgrade your systems today!
We all know the frustration of discovering that your fridge or freezer has failed due to an open door. With the AiroSensor 20-20-41 combined with the Magnetic Door Contact Set (both sold separately), you can say goodbye to this common problem.
With these two products, which should both be ordered; you can program alarms for open or improperly closed doors. This innovative solutionMore Product Information