Insect cell screened fetal bovine serum (FBS), U.S. Origin, Hyclone™
Leverandør: HyClone products (Cytiva)
SH30071.03I
SH30071.03IEA 9650 NOK
SH30071.03I
Insect cell screened fetal bovine serum (FBS), U.S. Origin, Hyclone™
Serum
Enhance cell culture growth with U.S. sourced Cytiva HyClone Insect Cell Screened Fetal Bovine Serum (FBS), filtered through serial 40 nm pore-size rated filters and identified through lots that offer exceptional performance with baculovirus expression vector systems (BEVS).
  • Optimises growth performance of insect cells
  • Sourced in the USA with complete traceability back to original source
  • Low in antibodies and high in growth factors
  • Virus panel testing according to 9 CFR 113,53
  • 40 nm filtration
With use of baculovirus expression vector systems (BEVS),More Product Information