Characterised fetal bovine serum (FBS), tetracycline screened, U.S. origin, HyClone™
Leverandør: HyClone products (Cytiva)
SH30071.03T
SH30071.03TEA 9650 NOK
SH30071.03T
Characterised fetal bovine serum (FBS), tetracycline screened, U.S. origin, HyClone™
Serum
Enhance growth of cells with U.S. sourced HyClone Characterized Fetal Bovine Serum (FBS).
  • HyClone characterized fetal bovine serum (FBS) is sourced in the USA. Complete traceability is provided back to the original source, as certified by the International Serum Industry Association (ISIA)
  • Additional traceability testing of premium HyClone fetal bovine serum is performed by our collaborator, Oritain
  • It is low in antibodies and high in growth factors to support robust growth and division of cells, including embryonic stem cells
HyClone characterizedMore Product Information