HyClone™ Dialysed Foetal Bovine Serum, US origin
Leverandør: HyClone products (Cytiva)
SH30079.02 SH30079.03 SH30079.02HI SH30079.01HI SH30079.03HI SH30079.01 SH30079.03IR25
HYCLSH30079.02EA 2400 NOK
HYCLSH30079.02 HYCLSH30079.03 HYCLSH30079.02HI HYCLSH30079.01HI HYCLSH30079.03HI HYCLSH30079.01 SH30079.03IR25
HyClone™ Dialysed Foetal Bovine Serum, US origin
Serum
Augment growth of cells with U.S.-sourced Cytiva HyClone Dialyzed Fetal Bovine Serum (FBS).
  • Depleted of small molecules (less than 10000 MW)
Filtered through sequential 100 nm (0.1 μm) pore-size rated filters.
Dialysed foetal bovine serum (FBS) is used in applications requiring serum depleted of small molecules (<10000 MW) and is filtered through filtered through three sequential 100 nm (0,1 µm) pore-size related filters. The diafiltration process reduces concentrations of low molecular weight components, such as nucleotides and amino acids,More Product Information