Fetal bovine serum (United States), Hyclone™, standard
Leverandør: HyClone products (Cytiva)
SH30088.03IH25
SH30088.03IH25EA 8025 NOK
SH30088.03IH25
Fetal bovine serum (United States), Hyclone™, standard
Serum
HyClone standard fetal bovine serum (FBS), U.S. origin is an economical alternative to characterised and defined FBS. Standard FBS is ideal for research applications.
  • Sourced in the USA with complete traceability back to original source
  • Low in antibodies and high in growth factors
  • Produced using true pool technology for uniformity and consistency from bottle to bottle
  • Filtered through three sequential 100 nm (0.1 µm) pore size-rated filters
  • Virus panel testing according to 9 CFR 113.53
Standard FBS is assayed for gamma globulin, alkaline phosphatase,More Product Information
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Lagringstemp Less than –10 °C