Flame photometer
Leverandør: SHERWOOD SCIENTIFIC INSTRUMENT
47542500
SHEW47542500EA 0 NOK
SHEW47542500
Flame photometer
Spektrofotometre og fotometre Flammefotometere
Designed to measure up to four elements: Sodium (Na), potassium (K) and calcium (Ca) using air/propane flame.
  • Fully automatic ignition and flame optimisation
  • Date and time of analysis printed at each calibration
  • Quality check and repeat samples identified on the printout
In 'Continuous' mode, it displays the current value on each channel: A reading can be printed at any time.
In 'Peak' mode, it can detect a stable reading following sample introduction for transfer to a printer or computer and the display is frozen until the next sample.