Villanueva Osteochrome bone stain
Leverandør: SIGMA ALDRICH MICROSCOPY
Danger
SHH0027-450ML
SHH0027-450MLEA 2600 NOK
SHH0027-450ML
Villanueva Osteochrome bone stain
Villanueva Osteochrome Bone stain
Useful for staining fresh, fixed, unembedded or plastic-embedded sections of bone. Villanueva osteochrome bone stain gives uniform and reproducible results. Useful in studying biopsy or postmortem tissue.