Dehydrated culture media, R-2A agar suitable for microbiology, NutriSelect® Plus
Leverandør: Merck
17209-500G
SIAL17209-500GEA 2680 NOK
SIAL17209-500G
Dehydrated culture media, R-2A agar suitable for microbiology, NutriSelect® Plus
Mikrobiologiske medier Media, dehydrert for mikrobiologi
R-2A agar is used for heterotrophic plate count of water samples using longer incubation periods.