Dehydrated culture media, china blue lactose agar
Leverandør: Merck
22520-500G
SIAL22520-500GEA 2890 NOK
SIAL22520-500G
Dehydrated culture media, china blue lactose agar
Mikrobiologiske medier Media, dehydrert for mikrobiologi
Selective medium for differentiating lactose fermenting and lactose non-fermenting microorganisms.