Dey-engley neutralising broth
Leverandør: Merck
D3435-500G
SIALD3435-500GEA 2800 NOK
SIALD3435-500G
Dey-engley neutralising broth
Mikrobiologiske medier Media, dehydrert for mikrobiologi
Dey-Engley neutralising broth is used in disinfectant testing where neutralisation of the antiseptics and disinfectants is important for determining bactericidal activity.
Components include casein enzymatic hydrolysate (5,00), yeast extract (2,50), dextrose (10,0), sodium thiosulphate (6,00), sodium thioglycollate (1,00), sodium bisulphite (2,50), lecithin (7,00), polysorbate (80 5,00) and bromocresol purple (0,02).