Temperature loggers, metal sheet and coil profiler
Leverandør: Industrial Physics
CX3080
SIMECX3080EA 64425 NOK
SIMECX3080
Temperature loggers, metal sheet and coil profiler
Dataloggere
The metal sheet and coil coating cure profiler monitors the temperature profile of the coil strip at various points across its width as it is fed through the oven.
  • Made of aluminium/stainless steel
  • Six measuring points (3-air and 3-surface)
  • Easy grab handle
Without stopping the line, the metal sheet and coil coating cure profiler can be put on the moving coil. It is not possible to mount this system on vertical coil production facilities.
Leveringsinformasjon: Set includes special bracket, CurveX 2 USB oven logger, insulation box CX2003 (max.More Product Information