Pocket torch
Leverandør: Industrial Physics
DI0035
SIMEDI0035EA 740 NOK
SIMEDI0035
Pocket torch
Lys Lommelykter
This powerful TQC aluminium LED pocket flashlight produces a very impressive 200 lumens and provides a projection distance of 250 m. It is suitable for applications in protective coatings, corrosion control, coating laboratories, paint production, surface finishing, powder coating, decorative coatings and building maintenance.
  • Dynamic switch
  • Shockproof casing
  • Four different modes: High beam, low beam, strobe mode, SOS signal mode
Weight: 73 g
Leveringsinformasjon: Supplied with a holster, a wriststrap, three AAA alkaline batteries and a gift box.