Concrete inspection hammer
Leverandør: Industrial Physics
LD0500
SIMELD0500EA 10675 NOK
SIMELD0500
Concrete inspection hammer
Beleggtester
Hammer used to test the quality of concrete, as well as other construction materials, for analysing resistance.
Sertifiseringer: EN 12504-2, ASTM, C805, BS 1881.