Density meter
Leverandør: Industrial Physics
LD0560
SIMELD0560EA 66850 NOK
SIMELD0560
Density meter
Densitetsmålere
This densimeter is designed to perform automatic calculation of density and compensation of water temperature.
  • Able to measure floating sample in water
  • Volume of solid sample can be measured
  • Values automatically displayed after the elapsed time