UV pocket flashlight
Leverandør: Industrial Physics
LD7290
SIMELD7290EA 1150 NOK
SIMELD7290
UV pocket flashlight
Lys UV-lamper
This UV pocket flashlight is designed to detect contaminations that react under UV illumination and can not be seen with naked eye such as some organic fats and alkaline contaminants. They are also suitable to inspect the cleanliness of steel prior to painting.
W×D: 32×126 mm
Leveringsinformasjon: Supplied with 3× AAA alkaline batteries.