Adhesion tester, PAThandy™
Leverandør: Industrial Physics
LD9200
SIMELD9200EA 79900 NOK
SIMELD9200
Adhesion tester, PAThandy™
Adhesjons testere
This instrument is an easy to use, lightweight hydraulic adhesion tester that applies an increasing level of pull off force to a dolly adhered to the surface under test simply by turning the handle. The adhesion test head has self adjusting legs, which ensure that the pull off force is always applied 90° to the coating even on uneven substrate surfaces. The self adjusting feature results in extremely accurate and reproducible adhesion test results.
  • Palm-held, light alloy pull-off tester
  • Case fits easily in the hand luggage (size of an A4 paper and only 80