Water penetration tester, acc. to Karsten
Leverandør: Industrial Physics
LI7500
SIMELI7500EA 5750 NOK
SIMELI7500
Water penetration tester, acc. to Karsten
Vanninntrenging testere
This Karsten tube penetration test is a simple test for measuring the degree or water penetration into building materials such as concrete, stone and plaster. The test kit consists of a glass tube filled with water, bonded to the test material with plastiline. Water pressure is then exerted on the surface. A graduated scale indicates, over time, the amount or water penetrated into the surface.
  • Tube material: Glass
  • Suitable for vertical and horizontal surfaces
Bestillingsinformasjon: The TQC Karsten Tube Penetration Test contains 3 tubes for horizontalMore Product Information