Concrete moisture tester, Tramex®
Leverandør: Industrial Physics
LI9011
SIMELI9011EA 9925 NOK
SIMELI9011
Concrete moisture tester, Tramex®
Analysatorer Fuktanalyse
Tramex is a non-destructive instrument for measuring moisture levels in concrete and sand-cement floors.
  • Measurements up to a few centimeters deep
  • Can be taken without damaging the surface
For use when laying parquet floors, floor coverings or floor coatings.