Concrete moisture analyser
Leverandør: Industrial Physics
LI9200
SIMELI9200EA 7175 NOK
SIMELI9200
Concrete moisture analyser
Analysatorer Fuktanalyse
This meter is a non-destructive moisture meter for concrete. By measuring the electrical impedance, the moisture content of concrete can be determined. Readings are taken by pressing the instrument against the surface.
  • Quick testing of large concrete floors or constructions
  • Automatic turn-off option
  • Four scales with normal and maximum 'HOLD' mode
The electrical impedance is measured by generating a low frequency electric field between the eight electrodes. Depending on the moisture content, the measurements are taken at a depth of several centimetres.
More Product Information
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Fuktighetsområde 5 − 40 %
Nøyaktighet ±0,5 %
Display Backlit monochrome (128×64 p) 61×33 mm
B×D×H 147×89×33 mm