Paint inspection gauge, Super Pig III
Leverandør: Industrial Physics
SP1100
SIMESP1100EA 13125 NOK
SIMESP1100
Paint inspection gauge, Super Pig III
Beleggtester
This inspection gauge is a destructive precision tool for inspection and thickness measurement, on single or multiple coats, on virtually all substrates, including wood, plastics, metals.
  • Titanium anodised aluminum
  • Three knife sizes and one crosscut knife in one holder
  • Ergonomic design with revolving system
  • Bright white LED ensure clear vision through the microscope
  • Small size eases use in corners
  • Engraved scaling (D-factor) for easy calculations
Observes and measures substrate and film defects. It is a very stable instrument, also the integratedMore Product Information