Indirect impact tester
Leverandør: Industrial Physics
SP1880
SIMESP1880EA 31950 NOK
SIMESP1880
Indirect impact tester
Beleggtester
Impact testers used to determine the impact resistivity and flexibility of materials.
  • Spirit level built-in for correct positioning
  • Dual scale instrument equipped with a special guidance
  • Distance between each impact is always according to the standards
Sertifiseringer: ISO 6272-2 and ASTM D2794