Linear abraser, Taber® 5750
Leverandør: Industrial Physics
TB0165
SIMETB0165EA 0 NOK
SIMETB0165
Linear abraser, Taber® 5750
Abrasjons testere
This linear wear resistance tester is designed to test virtually any size or shape specimen. Ideal for testing material properties of contoured surfaces and finished products.
  • Eleven stroke lengths (0,2" to 4,0")
  • Variable stroke speed from 2 to 75 cycles per minute
  • Preset stroke speed buttons for 2, 15, 25, 30, 40, 60 cycles per minute
  • Variable load from 350 to 2100 g with optional weight discs
  • Stainless Steel Wearaser Collet for use with vitrified or resilient Wearsers
  • Laser alignment guide
  • Programmable up to 999 999 cycles
The linear abraser uses aMore Product Information