Film applicators, multi-functional
Leverandør: Industrial Physics
VF1550
SIMEVF1550EA 8875 NOK
SIMEVF1550
Film applicators, multi-functional
Filmapplikatorer Paint/Ink Film Applicators
Multi-functional film applicator (Octoplex) with 8 pre-defined thickness sides for applying paint films of 25, 50, 75, 100, 125, 150, 175, 200 µm. Film width 60 mm. The high-grade stainless steel will not be affected by acid or base elements.