Temperature gradient plate for MFFT test
Leverandør: Industrial Physics
VF9700 VF9600
SIMEVF9700EA 0 NOK
SIMEVF9700 SIMEVF9600
Temperature gradient plate for MFFT test
Filmapplikatorer Sealing Film Applicators
Temperature gradient plate for determining minimum film forming temperature (MFFT) test. The instrument can be used to define the white point/glass transition temperature of dispersion materials, synthetic resins, enamels and the blocking power and stacking capability of coated papers, foils, prints.
  • Ground hard-chrome plated metal platen as to deposit the specimen
  • Displays nominal and actual value of hot and cold side
  • Electronic controller with digital display, temperature preselecting and overshoot protection
  • Integrated membrane drying unit with flow