Urine containers, SpecTainer™
Leverandør: Simport Scientific
C566-120CYS
SIMPC566-120CYSEA 2050 NOK
SIMPC566-120CYS
Urine containers, SpecTainer™
Diagnostiske systemer Clinical Sample Collection/Preparation Urine Sample Collection/Preparation
PP urine container with tamper evident PE screw cap, ideal for collecting, storing and transporting urine samples.
  • Tamper evident sealing ring for better sample protection
  • Ridges around base offer a better grip during opening and closing
For applications needing the utmost security where sample integrity is of high importance. A great feature of the Spectainer™ I is that it incorporates a unique tamper evident screw cap ensuring peace of mind during transport or storage situations where someone might have tampered with the specimen. For colour codingMore Product Information