Weighing pour-boats, disposable, anti-static, PS
Leverandør: Simport Scientific
D251-1 D251-2 D251-3
SIMPD251-1EA 385 NOK
SIMPD251-1 SIMPD251-2 SIMPD251-3
Weighing pour-boats, disposable, anti-static, PS
Skip Veieskip
Pour boats are made of antistatic polystyrene to facilitate dispensing of powdered and liquid materials. It can be also used to safely weigh static-affected materials.
  • Smooth, uniform and economical
  • Withstand temperatures up to 80 °C
  • Molded material used is thicker than conventional weighing dishes