Block mailer, paraffin
Leverandør: Simport Scientific
M477-6
SIMPM477-6EA 1775 NOK
SIMPM477-6
Block mailer, paraffin
Forsending og transport kontainere
PVC, translucent.
  • Sturdy, secure and easy to use
  • Easy viewing of contents
This mailer can be used for handling and shipping up to 6 blocks as needed and is suitable for all regular models of tissue and biopsy cassettes.