Microscope diamond slide writers
Leverandør: Simport Scientific
M795-1
SIMPM795-1EA 5000 NOK
SIMPM795-1
Microscope diamond slide writers
Penner Stylus and Scribers
The microscope diamond slide writer is compatible with a wide variety of slide labellers.
  • Consistent even coating of diamond dust
  • Stainless steel shaft
This diamond stylus is perfectly suited for the Thermo Scientific PrintMate and MicroWriter, RA Lamb, and TBS Shurmark microscope slide labellers. Manufactured with a consistent even coating of diamond dust.
The 2,35 mm shaft is made from stainless steel.