Microtube racks, UniRack™
Leverandør: Simport Scientific
S501-80 S500-80AS S500-80B S500-80G S500-80O S500-80P S500-80Y S500-80R
SIMPS501-80EA 505 NOK
SIMPS501-80 SIMPS500-80AS 212-0866 SIMPS500-80G SIMPS500-80O SIMPS500-80P SIMPS500-80Y SIMPS500-80R
Microtube racks, UniRack™
Stativer for rør Tube Racks
PP, reversible
  • Racks can be connected laterally to build up extended storage
  • Detachable handles for ease of transport and stacking of full racks
  • Detachable transparent lid available
Reversible racks holds up to 80 microtubes on one side or flip it over to hold up to 60 PCR tubes on the other.