Loading rack, RotoCycler™ 72 (consumables for Qiagen Rotor-Gene™ Q)
Leverandør: Simport Scientific
T319-4WS1
SIMPT319-4WS1EA 8050 NOK
SIMPT319-4WS1
Loading rack, RotoCycler™ 72 (consumables for Qiagen Rotor-Gene™ Q)
Stativer for rør Tube Racks
Solid aluminium loading rack for handling and insertion of caps on tubes.
Holds up to 18×0,1 ml tube strips (total of 72 tubes) and other cavities can hold larger reaction tubes. It should be simply place in the refrigerated area to keep reactions cool during setup.
All wells are numbered and colour coding is made possible by inserting a Capinsert™ in up to two locations.
Five hundred colour coding inserts of assorted colours are enclosed.