Screw cap with septum for microcentrifuge tubes, Micrewtube™
Leverandør: Simport Scientific
T347AQX
SIMPT347AQXEA 2275 NOK
SIMPT347AQX
Screw cap with septum for microcentrifuge tubes, Micrewtube™
Lokk Lokk til reagensglass
This screw cap incorporates a pierceable septum made of chemically resistant PTFE on the outside and silicone on the inside, both components being stable over a broad range of temperatures. The septum also acts as a silicone o-ring for better sample protection.
  • Made of polypropylene
  • Autoclavable
It is especially made to fit and be used with all Micrew™ microcentrifuge tubes. The cap is pierceable with pipette tips as well as with syringe needles.
Sertifiseringer: Microcentrifuge tubes and caps are certified RNase, DNase, DNA and Pyrogen free.