Tissue stainer, MSM/24
Leverandør: Slee
12000205
SLEE12000205EA 0 NOK
SLEE12000205
Tissue stainer, MSM/24
Fargetrau for objektglass
The MSM/24 carousel stainer with increased user safety with fume extraction efficiently eliminates hazardous fumes.
  • Integrated battery ensuring highest process
  • Safety even in environments with frequent power failures
Designed to meet the requirements for routine staining in histology and cytology laboratories.
Leveringsinformasjon: Supplied with operation manual, 24 plastic cuvettes, transport basket (plastic) with 30 standard size micro slides, and main cable.