Sterilin® Dippas® Milk Dippers
Leverandør: STERILIN
191 192 191BLUE 194IWBLUE 192BLUE
SLIN191EA 3275 NOK
SLIN191 SLIN192 SLIN191BLUE SLIN194IWBLUE SLIN192BLUE
Sterilin® Dippas® Milk Dippers
Prøvetakere Prøveøser
These sample dippers facilitate the sample collection and subsequent transportation to the laboratory in a single container with no risk of cross contamination.
  • Individually wrapped and gamma irradiated to ensure sterility
  • Available in either clear or blue PS for the food industry
  • Handle neatly snaps off after sample collection for ease of transportation
Sertifiseringer: EN 14245 and BS5213.
Leveringsinformasjon: Supplied with a 'flow-seal' screw cap for a leak tight seal.
NB: *Produktet/produktene på denne siden er utenfor omfanget av EU -direktivet 2019/904