DNA ladders, ultra low range, FastRuler
Leverandør: Thermo Fisher Scientific
SM1103
SM1103EA 895 NOK
SM1103
DNA ladders, ultra low range, FastRuler
Elektroforese reagenser Molekylvekt markører
FastRuler ultra low range DNA ladder is specifically designed for fast sizing and approximate quantification of double-stranded DNA in 48 well (or 96-well) high throughput gels, as well as in conventional agarose gels. The ladder is a mixture of five blunt-ended chromatography-purified individual DNA fragments that are easily resolved in a short separation distance (10 to 20 mm) after an 8 to 14 minutes run on the appropriate agarose gel.
  • Especially useful for electrophoresis of PCR products
  • Fast separation (8 to 14 min)
  • Short separation distance (10 to 20 mm)
  • Sharp bands, easy-to-remember fragment sizes and quantities
  • Ready to use – premixed with loading dye
  • Supplied with loading dye solution for sample DNA
FastRuler ultra low range DNA ladder contains dephosphorylated DNA fragments and is ideal for forward 5-end labeling with T4 polynucleotide kinase.