Tip boxes for Qualitix® macro tips
Leverandør: Socorex Isba
312.10ER
SOCO312.10EREA 325 NOK
SOCO312.10ER
Tip boxes for Qualitix® macro tips
Pipette Tip Boxes and Racks
Empty rack for 24×10 ml macrotips, autoclavable.
Leveringsinformasjon: 4/pack.