Repetitive pipette, Stepper™ 416
Leverandør: Socorex Isba
416.5000
SOCO416.5000EA 8200 NOK
SOCO416.5000
Repetitive pipette, Stepper™ 416
Pipetter Pipetter, repeterene stepper
Adjustable pipette intended for reliable dispensing and high performance results. Selected materials offer outstanding shock resistance and compact shape greatly limit internal contamination. The pipette provides up to 73 doses within a large selection of 53 different volumes.
  • Volumes range from 10 to 5000 µl
  • Unique trigger action (Activation with 4-finger instead of thumb) eliminates thumb fatigue
  • Setting knobs bear clear indication of volumes and number of aliquots
  • Only three positive displacement syringes, colour coded
  • Self-locking mechanism