Repetitive microdispensing pipette, Acura® 865
Leverandør: Socorex Isba
865.0050 865.0200 865.1000
SOCO865.0050EA 8500 NOK
SOCO865.0050 SOCO865.0200 SOCO865.1000
Repetitive microdispensing pipette, Acura® 865
Pipetter Pipetter, repeterene stepper
Handheld microdispenser intended for repeated distribution of microliter volumes. Three models cover the range from 5 to 1000 µl.
  • Excellent ergonomics
  • Ultra soft plunger activation, large volume display
  • Versatile feeding through bottle, tubing or syringe
  • Easy in-lab calibration
  • Autoclavable fully assembled at 121°C
  • Three-way valve system for automatic self-refilling
  • Carefully selected materials ensure durability and optimal resistance to aggressive media
Individually serial numbered and Q.C. certified
Leveringsinformasjon: Supplied with 90 cmMore Product Information