TriLink® Engineered RNase Inhibitor

Leverandør: TriLink BioTechnologies

E-0075-05 E-0075-006
SOLUE-0075-05EA 6325 NOK
SOLUE-0075-05 SOLUE-0075-006
TriLink® Engineered RNase Inhibitor
Nukleinsyre reagenser RNA Synthesis and Transcription

Engineered RNase Inhibitor is a protein to inhibit RNase activities to prevent RNA degradation in IVT.


  • Prevents degradation of synthesized RNA in vitro transcription (IVT)

  • Engineering improves resistance to oxidation compared to the human/porcine RNase inhibitors

  • Grade: Molecular biology


Engineered RNase Inhibitor binds to ribonucleases A, B, and C in a 1:1 stoichiometry with femtomolar affinity and renders them unable to bind or degrade RNA. Engineered RNase Inhibitor has significantly improved resistance to oxidation compared to the human/porcine RNase inhibitors and is stable at low DTT concentrations.


The provided 60 and 530 µg pack sizes contain approximately 3000 units and 26500 units of enzymes, respectively. One unit of the enzyme inhibits the activity of 5 ng of RNase A by 50%.


NB: Item will be delivered at or below –15 to –25 °C and should be stored at or below –15 to –25 °C.

Bestill nå

SPESIFIKASJONER

Environmentally Preferable
Applikasjon
Konsentrasjon (0,8 mg/ml, approximately 40 U/μl)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR